Anti-PTPRC Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 30-F11]

Supplier: R & D Systems
FAB114A
102880-206EA 399.14 USD
102880-206
Anti-PTPRC Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 30-F11]
Antibodies
Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone: 30-F11
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR