Anti-DLK1 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 211309]

Supplier: R & D Systems
FAB1144G-025 FAB1144G-100
102880-202EA 253.08 USD
102880-202 102880-204
Anti-DLK1 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 211309]
Antibodies
Type: Primary
Antigen: DLK1
Clonality: Monoclonal
Clone: 211309
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR