Anti-DLK1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 211309]

Supplier: R & D Systems
FAB1144A-100 FAB1144A-025
102880-198EA 308.03 USD
102880-198 102880-200
Anti-DLK1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 211309]
Antibodies
Type: Primary
Antigen: DLK1
Clonality: Monoclonal
Clone: 211309
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR