Anti-SIGLEC9 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 191240]

Supplier: R & D Systems
FAB1139P-025 FAB1139P-100
102880-194EA 202.46 USD
102880-194 102880-196
Anti-SIGLEC9 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 191240]
Antibodies
Type: Primary
Antigen: SIGLEC9
Clonality: Monoclonal
Clone: 191240
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR