Anti-SIGLEC9 Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: 191240]

Supplier: R & D Systems
FAB1139N-025 FAB1139N-100
102881-078EA 193.78 USD
102881-078 102880-192
Anti-SIGLEC9 Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: 191240]
Antibodies
Type: Primary
Antigen: SIGLEC9
Clonality: Monoclonal
Clone: 191240
Conjugation: Alexa Fluor® 700
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR