Anti-SIGLEC9 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 191240]

Supplier: R & D Systems
FAB1139G-100 FAB1139G-025
102881-076EA 561.11 USD
102881-076 102881-074
Anti-SIGLEC9 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 191240]
Antibodies
Type: Primary
Antigen: SIGLEC9
Clonality: Monoclonal
Clone: 191240
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR