Anti-SIGLEC9 Mouse Monoclonal Antibody (Fluorescein) [clone: 191240]

Supplier: R & D Systems
FAB1139F-025 FAB1139F-100
102881-070EA 193.78 USD
102881-070 102881-072
Anti-SIGLEC9 Mouse Monoclonal Antibody (Fluorescein) [clone: 191240]
Antibodies
Type: Primary
Antigen: SIGLEC9
Clonality: Monoclonal
Clone: 191240
Conjugation: Fluorescein
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR