Anti-SIGLEC7 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 194211]

Supplier: R & D Systems
FAB11381A-100 FAB11381A-025
102881-052EA 608.83 USD
102881-052 102881-050
Anti-SIGLEC7 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 194211]
Antibodies
Type: Primary
Antigen: SIGLEC7
Clonality: Monoclonal
Clone: 194211
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR