Anti-CD33 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 6C5/2]

Supplier: R & D Systems
FAB1137P-025 FAB1137P-100
102881-046EA 160.52 USD
102881-046 102881-048
Anti-CD33 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 6C5/2]
Antibodies
Type: Primary
Antigen: CD33
Clonality: Monoclonal
Clone: 6C5/2
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR