Anti-ERBB4 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 182818]

Supplier: R & D Systems
FAB11311P
220016-955EA 543.75 USD
220016-955
Anti-ERBB4 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 182818]
Antibodies
Type: Primary
Antigen: ERBB4
Clonality: Monoclonal
Clone: 182818
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR