Anti-ERBB4 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 182818]

Supplier: R & D Systems
FAB11311A
220016-954EA 594.37 USD
220016-954
Anti-ERBB4 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 182818]
Antibodies
Type: Primary
Antigen: ERBB4
Clonality: Monoclonal
Clone: 182818
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR