Anti-ERBB2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 191924]

Supplier: R & D Systems
FAB1129P
220016-953EA 566.89 USD
220016-953
Anti-ERBB2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 191924]
Antibodies
Type: Primary
Antigen: ERBB2
Clonality: Monoclonal
Clone: 191924
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR