Anti-LY76 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: TER-119]

Supplier: R & D Systems
FAB1125P
220016-951EA 513.38 USD
220016-951
Anti-LY76 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: TER-119]
Antibodies
Type: Primary
Antigen: LY76
Clonality: Monoclonal
Clone: TER-119
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR