Anti-FZD1 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 162531]

Supplier: R & D Systems
FAB11201A
220016-943EA 561.11 USD
220016-943
Anti-FZD1 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 162531]
Antibodies
Type: Primary
Antigen Symbol: FZD1
Antigen Name: frizzled class receptor 1
Antigen Synonyms: -
Clonality: Monoclonal
Clone: 162531
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR