Anti-IgG1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 97924]

Supplier: R & D Systems
FAB110A
MSPP-FAB110AEA 286.34 USD
MSPP-FAB110A
Anti-IgG1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 97924]
Antibodies
Type: Primary
Antigen: IgG1
Clonality: Monoclonal
Clone: 97924
Conjugation: APC (Allophycocyanin)
Epitope: F(c)
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR