Anti-EGFR Rat Monoclonal Antibody (Alexa Fluor® 488) [clone: 423103]

Supplier: R & D Systems
FAB10951G
102881-022EA 561.11 USD
102881-022
Anti-EGFR Rat Monoclonal Antibody (Alexa Fluor® 488) [clone: 423103]
Antibodies
Type: Primary
Antigen: EGFR
Clonality: Monoclonal
Clone: 423103
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR