Anti-IFNAR2 Goat Polyclonal Antibody (Fluorescein)

Supplier: R & D Systems
FAB1083F
102881-018EA 389.02 USD
102881-018
Anti-IFNAR2 Goat Polyclonal Antibody (Fluorescein)
Antibodies
Type: Primary
Antigen: IFNAR2
Clonality: Polyclonal
Clone:
Conjugation: Fluorescein
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR