Anti-KLRC1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 131411]

Supplier: R & D Systems
FAB1059P-025 FAB1059P-100
102880-184EA 655.11 USD
102880-184 102880-182
Anti-KLRC1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 131411]
Antibodies
Type: Primary
Antigen: KLRC1
Clonality: Monoclonal
Clone: 131411
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR