Anti-KLRC1 Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: 131411]

Supplier: R & D Systems
FAB1059N-025 FAB1059N-100
102880-178EA 241.51 USD
102880-178 102880-180
Anti-KLRC1 Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: 131411]
Antibodies
Type: Primary
Antigen: KLRC1
Clonality: Monoclonal
Clone: 131411
Conjugation: Alexa Fluor® 700
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR