Anti-KLRD1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 131412]

Supplier: R & D Systems
FAB1058P
220016-927EA 420.83 USD
220016-927
Anti-KLRD1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 131412]
Antibodies
Type: Primary
Antigen: KLRD1
Clonality: Monoclonal
Clone: 131412
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR