Anti-S100 A12 Goat Polyclonal Antibody (Alexa Fluor® 488)

Supplier: R & D Systems
FAB1052G
102880-160EA 561.11 USD
102880-160
Anti-S100 A12 Goat Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
Type: Primary
Antigen: S100 A12
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR