Anti-TNFRSF13B Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 166010]

Supplier: R & D Systems
FAB1041P
220016-923EA 513.38 USD
220016-923
Anti-TNFRSF13B Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 166010]
Antibodies
Type: Primary
Antigen: TNFRSF13B
Clonality: Monoclonal
Clone: 166010
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR