Anti-TNFRSF13B Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 166010]

Supplier: R & D Systems
FAB1041A
102881-014EA 535.08 USD
102881-014
Anti-TNFRSF13B Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 166010]
Antibodies
Type: Primary
Antigen: TNFRSF13B
Clonality: Monoclonal
Clone: 166010
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR