Anti-IL17RB Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 752101]

Supplier: R & D Systems
FAB10402P-100 FAB10402P-025
102881-012EA 513.38 USD
102881-012 102881-010
Anti-IL17RB Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 752101]
Antibodies
Type: Primary
Antigen: IL17RB
Clonality: Monoclonal
Clone: 752101
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR