Anti-IL17RB Rat Monoclonal Antibody (Alexa Fluor® 488) [clone: 752101]

Supplier: R & D Systems
FAB10402G-025 FAB10402G-100
102881-006EA 527.85 USD
102881-006 102881-004
Anti-IL17RB Rat Monoclonal Antibody (Alexa Fluor® 488) [clone: 752101]
Antibodies
Type: Primary
Antigen: IL17RB
Clonality: Monoclonal
Clone: 752101
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR