Anti-TNFSF8 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 116614]

Supplier: R & D Systems
FAB1028P
220016-916EA 517.72 USD
220016-916
Anti-TNFSF8 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 116614]
Antibodies
Type: Primary
Antigen: TNFSF8
Clonality: Monoclonal
Clone: 116614
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR