Anti-TNFSF8 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 116614]

Supplier: R & D Systems
FAB1028A
220016-915EA 588.58 USD
220016-915
Anti-TNFSF8 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 116614]
Antibodies
Type: Primary
Antigen: TNFSF8
Clonality: Monoclonal
Clone: 116614
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR