Anti-CD276 Mouse Monoclonal Antibody (Fluorescein) [clone: 185504]

Supplier: R & D Systems
FAB1027F
220016-913EA 420.83 USD
220016-913
Anti-CD276 Mouse Monoclonal Antibody (Fluorescein) [clone: 185504]
Antibodies
Type: Primary
Antigen: CD276
Clonality: Monoclonal
Clone: 185504
Conjugation: Fluorescein
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR