Anti-IL2RA Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 24212]

Supplier: R & D Systems
FAB1020G-100 FAB1020G-025
102881-000EA 404.92 USD
102881-000 102879-902
Anti-IL2RA Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 24212]
Antibodies
Type: Primary
Antigen: IL2RA
Clonality: Monoclonal
Clone: 24212
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR