Anti-CD274 Goat Polyclonal Antibody (APC (Allophycocyanin))

Supplier: R & D Systems
FAB1019A-025
MSPP-FAB1019AEA 274.77 USD
MSPP-FAB1019A
Anti-CD274 Goat Polyclonal Antibody (APC (Allophycocyanin))
Antibodies
Type: Primary
Antigen: CD274
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Goat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR