Anti-MPL Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 167639]

Supplier: R & D Systems
FAB1016P
102879-896EA 513.38 USD
102879-896
Anti-MPL Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 167639]
Antibodies
Type: Primary
Antigen: MPL
Clonality: Monoclonal
Clone: 167639
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR