Anti-MPL Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 167639]

Supplier: R & D Systems
FAB1016A
220016-902EA 577.02 USD
220016-902
Anti-MPL Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 167639]
Antibodies
Type: Primary
Antigen: MPL
Clonality: Monoclonal
Clone: 167639
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR