Anti-CD3E Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: UCHT1]

Supplier: R & D Systems
FAB100P-025 FAB100P-100
102879-894EA 389.02 USD
102879-894 102879-892
Anti-CD3E Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: UCHT1]
Antibodies
Type: Primary
Antigen: CD3E
Clonality: Monoclonal
Clone: UCHT1
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR