Anti-CD3E Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: UCHT1]

Supplier: R & D Systems
FAB100N-025 FAB100N-100
102879-890EA 446.86 USD
102879-890 102879-888
Anti-CD3E Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: UCHT1]
Antibodies
Type: Primary
Antigen: CD3E
Clonality: Monoclonal
Clone: UCHT1
Conjugation: Alexa Fluor® 700
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR