Anti-CD3E Mouse Monoclonal Antibody (Fluorescein) [clone: UCHT1]

Supplier: R & D Systems
FAB100F-100 FAB100F-025
102879-884EA 177.88 USD
102879-884 102879-886
Anti-CD3E Mouse Monoclonal Antibody (Fluorescein) [clone: UCHT1]
Antibodies
Type: Primary
Antigen: CD3E
Clonality: Monoclonal
Clone: UCHT1
Conjugation: Fluorescein
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR