Anti-CD3E Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: UCHT1]

Supplier: R & D Systems
FAB100A-100 FAB100A-025
102879-878EA 458.43 USD
102879-878 102879-876
Anti-CD3E Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: UCHT1]
Antibodies
Type: Primary
Antigen: CD3E
Clonality: Monoclonal
Clone: UCHT1
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR