Anti-IL1RL1 Rat Monoclonal Antibody (Alexa Fluor® 700) [clone: 245707]

Supplier: R & D Systems
FAB10041N-100 FAB10041N-025
102879-870EA 501.82 USD
102879-870 102879-868
Anti-IL1RL1 Rat Monoclonal Antibody (Alexa Fluor® 700) [clone: 245707]
Antibodies
Type: Primary
Antigen: IL1RL1
Clonality: Monoclonal
Clone: 245707
Conjugation: Alexa Fluor® 700
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR