Anti-CDH5 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 162709]

Supplier: R & D Systems
FAB1002A
102879-860EA 561.11 USD
102879-860
Anti-CDH5 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 162709]
Antibodies
Type: Primary
Antigen: CDH5
Clonality: Monoclonal
Clone: 162709
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR