Anti-LTB4R Mouse Monoclonal Antibody (Fluorescein) [clone: 203/14F11]

Supplier: R & D Systems
FAB099F
220016-891EA 404.92 USD
220016-891
Anti-LTB4R Mouse Monoclonal Antibody (Fluorescein) [clone: 203/14F11]
Antibodies
Type: Primary
Antigen: LTB4R
Clonality: Monoclonal
Clone: 203/14F11
Conjugation: Fluorescein
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR