Anti-IgG2 beta Rat Monoclonal Antibody [clone: 332723] (Fluorescein)

Supplier: R & D Systems
F0134
102879-716EA 261.75 USD
102879-716
Anti-IgG2 beta Rat Monoclonal Antibody [clone: 332723] (Fluorescein)
Antibodies
Type: Secondary
Antigen: IgG2 beta
Clonality: Monoclonal
Clone: 332723
Conjugation: Fluorescein
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR