Anti-IgG2 beta Rat Monoclonal Antibody [clone: 332723] (PE (Phycoerythrin))

Supplier: R & D Systems
F0132
102879-712EA 326.83 USD
102879-712
Anti-IgG2 beta Rat Monoclonal Antibody [clone: 332723] (PE (Phycoerythrin))
Antibodies
Type: Secondary
Antigen: IgG2 beta
Clonality: Monoclonal
Clone: 332723
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR