Anti-IgG2a Rat Monoclonal Antibody [clone: 344701] (PerCP (Peridinin-Chlorophyll Protein Complex))

Supplier: R & D Systems
F0131
102879-710EA 344.18 USD
102879-710
Anti-IgG2a Rat Monoclonal Antibody [clone: 344701] (PerCP (Peridinin-Chlorophyll Protein Complex))
Antibodies
Type: Secondary
Antigen: IgG2a
Clonality: Monoclonal
Clone: 344701
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR