Anti-IgG2a Rat Monoclonal Antibody [clone: 344701] (APC (Allophycocyanin))

Supplier: R & D Systems
F0130
102879-708EA 403.48 USD
102879-708
Anti-IgG2a Rat Monoclonal Antibody [clone: 344701] (APC (Allophycocyanin))
Antibodies
Type: Secondary
Antigen: IgG2a
Clonality: Monoclonal
Clone: 344701
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR