Anti-IgG Donkey Polyclonal Antibody (APC (Allophycocyanin))

Supplier: R & D Systems
F0127
102879-702EA 403.48 USD
102879-702
Anti-IgG Donkey Polyclonal Antibody (APC (Allophycocyanin))
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: Heavy and Light chain
Host: Donkey
Isotype: IgG
Reactivity: Sheep
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR