Anti-IgG Donkey Polyclonal Antibody (Fluorescein)

Supplier: R & D Systems
F0125
102879-698EA 261.75 USD
102879-698
Anti-IgG Donkey Polyclonal Antibody (Fluorescein)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Fluorescein
Epitope: Heavy and Light chain
Host: Donkey
Isotype: IgG
Reactivity: Sheep
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR