Anti-Hamster IgG Mouse Monoclonal Antibody [clone: MAH1.12] (APC (Allophycocyanin))

Supplier: R & D Systems
F0121
220016-886EA 403.48 USD
220016-886
Anti-Hamster IgG Mouse Monoclonal Antibody [clone: MAH1.12] (APC (Allophycocyanin))
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Monoclonal
Clone: MAH1.12
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Hamster
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR