Anti-IgM Goat Polyclonal Antibody (APC (Allophycocyanin))

Supplier: R & D Systems
F0117
220016-882EA 403.48 USD
220016-882
Anti-IgM Goat Polyclonal Antibody (APC (Allophycocyanin))
Antibodies
Type: Secondary
Antigen: IgM
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR