Anti-IgM Goat Polyclonal Antibody (PE (Phycoerythrin))

Supplier: R & D Systems
F0116
220016-881EA 326.83 USD
220016-881
Anti-IgM Goat Polyclonal Antibody (PE (Phycoerythrin))
Antibodies
Type: Secondary
Antigen: IgM
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR