Anti-IgG Goat Polyclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex))

Supplier: R & D Systems
F0115
220016-880EA 326.83 USD
220016-880
Anti-IgG Goat Polyclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex))
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)
Epitope: Heavy and Light Chain
Host: Goat
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR