Anti-IgG Goat Polyclonal Antibody (APC (Allophycocyanin))

Supplier: R & D Systems
F0111
220016-876EA 403.48 USD
220016-876
Anti-IgG Goat Polyclonal Antibody (APC (Allophycocyanin))
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR